Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZKSCAN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318082.1 | NP_001305011.1 | 7 | 4115 | ||
NM_001318083.1 | NP_001305012.1 | 5 | 3834 | |||
NM_001318084.1 | NP_001305013.1 | 5 | 3827 | |||
NM_014569.3 | NP_055384.1 | 7 | 4174 | |||
NM_145102.3 | NP_659570.1 | 7 | 4053 | |||
XM_011515999.2 | XP_011514301.1 | 6 | 3714 | |||
XM_017011918.1 | XP_016867407.1 | 9 | 4512 | |||
XM_017011919.1 | XP_016867408.1 | 8 | 4555 | |||
XM_017011920.1 | XP_016867409.1 | 6 | 4717 | |||
XM_017011921.1 | XP_016867410.1 | 7 | 4483 | |||
XM_017011922.1 | XP_016867411.1 | 7 | 4142 | |||
XM_017011923.1 | XP_016867412.1 | 7 | 4293 | |||
XM_017011924.1 | XP_016867413.1 | 5 | 3955 | |||
XM_017011925.1 | XP_016867414.1 | 6 | 3659 | |||
XM_017011926.1 | XP_016867415.1 | 6 | 4077 | |||
XM_017011927.1 | XP_016867416.1 | 6 | 4009 | |||
GenBank mRNA | AB023232.1 | - | 7 | 4115 | ||
AF170025.1 | - | 7 | 4027 | |||
BX648490.1 | - | 5 | 4580 |