Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271620.2 | NP_001258549.1 | 3 | 311 | ||
NM_015069.4 | NP_055884.2 | 3 | 201 | |||
XM_005255856.4 | XP_005255913.1 | 3 | 914 | |||
XM_006721171.3 | XP_006721234.1 | 6 | 643 | |||
XM_017023076.1 | XP_016878565.1 | 3 | 928 | |||
XM_017023077.1 | XP_016878566.1 | 3 | 277 | |||
XM_017023078.1 | XP_016878567.1 | 2 | 250 | |||
GenBank mRNA | AK314742.1 | - | 3 | 312 | ||
BC112315.1 | - | 3 | 203 | 8327783 | ||
BC112317.1 | - | 3 | 203 | 8327785 |