Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF337 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290261.1 | NP_001277190.1 | 4 | 1099 | ||
NM_015655.3 | NP_056470.1 | 5 | 772 | |||
XM_005260702.3 | XP_005260759.1 | 4 | 588 | |||
XM_006723558.3 | XP_006723621.1 | 5 | 1775 | |||
XM_011529219.2 | XP_011527521.1 | 5 | 895 | |||
XM_017027802.1 | XP_016883291.1 | 3 | 2670 | |||
XM_017027803.1 | XP_016883292.1 | 4 | 1679 | |||
XM_017027804.1 | XP_016883293.1 | 2 | 448 | |||
GenBank mRNA | AK056184.1 | - | 5 | 683 | ||
AK056466.1 | - | 1 | 295 | |||
AK125455.1 | - | 4 | 1102 | |||
AK291235.1 | - | 5 | 709 | |||
AK299811.1 | - | 3 | 774 | |||
BC021298.2 | - | 5 | 701 | 4551314 |