Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002483.6 | NP_002474.4 | 2 | 274 | ||
XM_011526990.1 | XP_011525292.1 | 2 | 373 | |||
GenBank mRNA | AK312542.1 | - | 2 | 259 | ||
BC005008.1 | - | 2 | 195 | 3640231 | ||
BT009774.1 | - | 2 | 155 | |||
DA442875.1 | - | 2 | 375 | |||
M18216.1 | - | 2 | 201 | |||
M18728.1 | - | 2 | 205 | |||
M29541.1 | - | 2 | 248 |