Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF691 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242739.1 | NP_001229668.1 | 4 | 880 | ||
NM_015911.3 | NP_056995.1 | 2 | 579 | |||
XM_006710671.3 | XP_006710734.1 | 3 | 696 | |||
XM_017001400.1 | XP_016856889.1 | 3 | 1025 | |||
XM_017001401.1 | XP_016856890.1 | 2 | 3562 | |||
XM_017001402.1 | XP_016856891.1 | 3 | 2026 | |||
XM_017001403.1 | XP_016856892.1 | 2 | 4562 | |||
GenBank mRNA | AF131739.1 | - | 2 | 543 | ||
AK000938.1 | - | 2 | 682 | |||
AK296201.1 | - | 3 | 968 | |||
BC000157.2 | - | 2 | 622 | 2901198 | ||
CR457146.1 | - | 1 | 485 |