Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016270.3 | NP_057354.1 | 3 | 1382 | ||
GenBank mRNA | AF134053.1 | - | 3 | 1284 | ||
AF205849.1 | - | 3 | 1368 | |||
AK225820.1 | - | 3 | 1381 | |||
BC071983.1 | - | 3 | 1371 | 5480246 | ||
BQ581981.1 | - | 2 | 418 |