Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF571 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290314.1 | NP_001277243.1 | 5 | 463 | ||
NM_001321272.1 | NP_001308201.1 | 4 | 971 | |||
NM_016536.4 | NP_057620.3 | 4 | 363 | |||
XM_011527024.2 | XP_011525326.1 | 4 | 445 | |||
XM_017026855.1 | XP_016882344.1 | 4 | 1296 | |||
XM_017026856.1 | XP_016882345.1 | 4 | 876 | |||
XM_017026857.1 | XP_016882346.1 | 4 | 1009 | |||
XM_017026858.1 | XP_016882347.1 | 4 | 378 | |||
XM_017026859.1 | XP_016882348.1 | 4 | 815 | |||
XM_017026860.1 | XP_016882349.1 | 3 | 274 | |||
GenBank mRNA | AF161544.2 | - | 4 | 344 | ||
AK291413.1 | - | 5 | 432 | |||
BC040591.1 | - | 4 | 971 | 5269760 | ||
BC113870.1 | - | 4 | 338 | 40036719 | ||
BC114479.1 | - | 4 | 342 | 40036718 | ||
BP312315.1 | - | 4 | 363 | |||
BX537401.1 | - | 4 | 362 |