Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173487.1 | NP_001166958.1 | 4 | 414 | ||
NM_001173488.1 | NP_001166959.1 | 2 | 800 | |||
NM_017544.3 | NP_060014.2 | 3 | 316 | |||
XM_011531365.2 | XP_011529667.1 | 4 | 946 | |||
GenBank mRNA | AJ011812.2 | - | 2 | 800 | ||
AL539002.3 | - | 4 | 343 | |||
AY208891.1 | - | 3 | 316 | |||
BC040379.1 | - | 3 | 298 | 4777539 | ||
BC047878.2 | - | 3 | 283 | 5228666 | ||
BC068514.1 | - | 3 | 294 | 5299667 | ||
EF158333.1 | - | 2 | 800 | |||
EF165090.1 | - | 2 | 540 | |||
EF165091.1 | - | 2 | 619 | |||
EF165093.1 | - | 2 | 690 | |||
EF216688.1 | - | 2 | 800 | |||
EF216689.1 | - | 2 | 800 |