Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017643.2 | NP_060113.2 | 10 | 1389 | ||
XM_005257467.4 | XP_005257524.1 | 9 | 1334 | |||
XM_005257468.4 | XP_005257525.1 | 9 | 1327 | |||
XM_011524922.2 | XP_011523224.1 | 9 | 1336 | |||
XM_011524923.2 | XP_011523225.1 | 9 | 1333 | |||
XM_011524924.2 | XP_011523226.1 | 9 | 1329 | |||
XM_011524925.1 | XP_011523227.1 | 10 | 1450 | |||
XM_011524926.2 | XP_011523228.1 | 10 | 1554 | |||
XM_011524928.2 | XP_011523230.1 | 9 | 1491 | |||
XM_011524929.2 | XP_011523231.1 | 10 | 1405 | |||
XM_011524930.1 | XP_011523232.1 | 9 | 1127 | |||
XM_011524931.2 | XP_011523233.1 | 8 | 961 | |||
XM_011524933.2 | XP_011523235.1 | 11 | 1511 | |||
XM_011524934.2 | XP_011523236.1 | 9 | 1335 | |||
XM_011524935.2 | XP_011523237.1 | 9 | 1328 | |||
XM_011524936.1 | XP_011523238.1 | 9 | 1127 | |||
XM_017024782.1 | XP_016880271.1 | 10 | 1701 | |||
GenBank mRNA | AK000062.1 | - | 6 | 721 | ||
AK124061.1 | - | 9 | 1127 | |||
BC034364.1 | - | 10 | 1360 | |||
BC101736.1 | - | 5 | 650 | 8069242 |