Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017643.2 | NP_060113.2 | 12 | 1526 | ||
XM_005257467.4 | XP_005257524.1 | 11 | 1471 | |||
XM_005257468.4 | XP_005257525.1 | 11 | 1464 | |||
XM_011524922.2 | XP_011523224.1 | 11 | 1473 | |||
XM_011524923.2 | XP_011523225.1 | 11 | 1470 | |||
XM_011524924.2 | XP_011523226.1 | 11 | 1466 | |||
XM_011524925.1 | XP_011523227.1 | 12 | 1587 | |||
XM_011524926.2 | XP_011523228.1 | 12 | 1691 | |||
XM_011524928.2 | XP_011523230.1 | 11 | 1628 | |||
XM_011524929.2 | XP_011523231.1 | 12 | 1542 | |||
XM_011524930.1 | XP_011523232.1 | 11 | 1264 | |||
XM_011524931.2 | XP_011523233.1 | 10 | 1098 | |||
XM_011524933.2 | XP_011523235.1 | 13 | 1648 | |||
XM_011524934.2 | XP_011523236.1 | 11 | 1472 | |||
XM_011524935.2 | XP_011523237.1 | 11 | 1465 | |||
XM_011524936.1 | XP_011523238.1 | 11 | 1264 | |||
XM_017024782.1 | XP_016880271.1 | 12 | 1838 | |||
GenBank mRNA | AK000062.1 | - | 8 | 858 | ||
AK124061.1 | - | 11 | 1264 | |||
BC034364.1 | - | 12 | 1497 | |||
BC101736.1 | - | 7 | 787 | 8069242 |