Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304496.1 | NP_001291425.1 | 3 | 351 | ||
NM_018073.7 | NP_060543.5 | 4 | 834 | |||
GenBank mRNA | AF360739.1 | - | 4 | 787 | ||
AF439153.1 | - | 3 | 544 | |||
AK001231.1 | - | 1 | 39 | |||
AK056523.1 | - | 4 | 796 | |||
AK291166.1 | - | 4 | 746 | |||
AK301120.1 | - | 3 | 234 | |||
AK310888.1 | - | 4 | 743 | |||
AL832730.1 | - | 4 | 784 | |||
AY005802.1 | - | 3 | 220 | |||
BC075058.2 | - | 3 | 568 | 30915367 | ||
BC109063.2 | - | 4 | 753 | 40033795 | ||
DA833140.1 | - | 3 | 234 |