Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042414.2 | NP_001035879.1 | 3 | 564 | ||
NR_003272.2 | 2 | 689 | ||||
NR_044998.1 | 2 | 689 | ||||
XM_006719844.3 | XP_006719907.1 | 2 | 717 | |||
XM_011535137.1 | XP_011533439.1 | 2 | 676 | |||
XM_011535138.2 | XP_011533440.1 | 2 | 676 | |||
XM_011535139.1 | XP_011533441.1 | 2 | 676 | |||
XM_011535140.1 | XP_011533442.1 | 2 | 676 | |||
XM_011535141.2 | XP_011533443.1 | 2 | 676 | |||
XM_011535142.2 | XP_011533444.1 | 2 | 676 | |||
XM_017020650.1 | XP_016876139.1 | 2 | 676 | |||
GenBank mRNA | AF448795.1 | - | 2 | 427 | ||
AF449627.1 | - | 2 | 427 | |||
AK001817.1 | - | 2 | 612 | |||
AK301554.1 | - | 4 | 389 | |||
BC014184.2 | - | 2 | 582 | 4623069 | ||
CR457272.1 | - | 2 | 319 | |||
DC396174.1 | - | 3 | 564 |