Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243116.1 | NP_001230045.1 | 3 | 430 | ||
NM_002503.4 | NP_002494.2 | 3 | 672 | |||
NR_040515.1 | 3 | 616 | ||||
XM_006723226.2 | XP_006723289.2 | 3 | 608 | |||
XM_006723227.3 | XP_006723290.3 | 3 | 571 | |||
GenBank mRNA | AB209784.1 | - | 3 | 638 | ||
AK057862.1 | - | 3 | 567 | |||
AK290569.1 | - | 3 | 623 | |||
BC007197.1 | - | 1 | 272 | |||
BC015528.1 | - | 3 | 603 | 3903794 | ||
BI520169.1 | - | 3 | 430 | |||
BM449613.1 | - | 3 | 627 | |||
BQ440907.1 | - | 3 | 404 | |||
BT006743.1 | - | 3 | 569 |