Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDCA7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127370.2 | NP_001120842.1 | 5 | 693 | ||
NM_001127371.2 | NP_001120843.1 | 3 | 501 | |||
NM_018719.4 | NP_061189.2 | 4 | 639 | |||
GenBank mRNA | AK022955.1 | - | 4 | 614 | ||
AK095965.1 | - | 5 | 647 | |||
AK292490.1 | - | 4 | 614 | |||
AK300781.1 | - | 3 | 461 | |||
AY161168.1 | - | 4 | 516 | |||
AY161169.1 | - | 4 | 519 | |||
BC009352.2 | - | 4 | 531 | 4139111 | ||
BC014630.2 | - | 3 | 483 | |||
BC025242.1 | - | 4 | 593 | 4842169 | ||
BC032576.1 | - | 4 | 604 | 5491780 | ||
BC039823.1 | - | 4 | 603 | 6057601 | ||
BX422040.2 | - | 3 | 449 | |||
DC331792.1 | - | 3 | 501 |