Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019896.2 | NP_063949.2 | 2, 3 | 327 | ||
GenBank mRNA | AI815728.1 | - | 2, 3 | 354 | ||
AY034104.1 | - | 1, 2 | 68 | |||
BC031331.1 | - | 2, 3 | 322 | 5443810 | ||
BU597500.1 | - | 334 | ||||
CA454961.1 | - | 2, 3 | 269 |