Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276378.1 | NP_001263307.1 | 12 | 2588 | ||
NM_001276379.1 | NP_001263308.1 | 10 | 2359 | |||
NM_020347.3 | NP_065080.1 | 10 | 2307 | |||
NR_075080.1 | 9 | 2182 | ||||
XM_006713207.2 | XP_006713270.2 | 9 | 2188 | |||
XM_011533838.2 | XP_011532140.1 | 10 | 2231 | |||
XM_017006644.1 | XP_016862133.1 | 13 | 2945 | |||
XM_017006645.1 | XP_016862134.1 | 12 | 2820 | |||
GenBank mRNA | AJ297351.1 | - | 10 | 2264 | ||
AK027165.1 | - | 1 | 935 | |||
AK129805.1 | - | 1 | 351 | |||
BC042483.1 | - | 10 | 2267 | 4831113 | ||
BX640604.1 | - | 10 | 2289 |