Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF670-ZNF695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037894.1 | 7 | 967 |
Gene Symbol: ZNF695 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204221.1 | NP_001191150.1 | 6 | 802 | ||
NR_037892.1 | 6 | 806 | ||||
GenBank mRNA | BC023527.2 | - | 6 | 714 | ||
BC041082.1 | - | 6 | 739 | |||
BC055096.1 | - | 5 | 635 | |||
BE743952.1 | - | 5 | 545 |