Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190791.1 | NP_001177720.1 | 6 | 850 | ||
NM_020933.4 | NP_065984.3 | 7 | 946 | |||
NR_102435.1 | 7 | 1092 | ||||
GenBank mRNA | AB046808.1 | - | 6 | 723 | ||
AF148135.1 | - | 6 | 833 | |||
AF275255.1 | - | 7 | 929 | |||
AF307096.1 | - | 4 | 1067 | |||
AF307097.1 | - | 5 | 1165 | |||
AK056570.1 | - | 7 | 929 | |||
AK304871.1 | - | 6 | 661 | |||
AK314560.1 | - | 7 | 929 | |||
AL834126.2 | - | 5 | 2021 | |||
BC078154.1 | - | 7 | 916 | 6060418 |