Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF529 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145649.1 | NP_001139121.1 | 6 | 672 | ||
NM_001145650.1 | NP_001139122.1 | 5 | 612 | |||
NM_001321351.1 | NP_001308280.1 | 6 | 691 | |||
NM_020951.4 | NP_066002.3 | 5 | 590 | |||
XM_006723302.3 | XP_006723365.1 | 5 | 464 | |||
XM_006723304.2 | XP_006723367.1 | 5 | 590 | |||
XM_011527164.2 | XP_011525466.1 | 6 | 607 | |||
XM_011527165.2 | XP_011525467.1 | 4 | 457 | |||
XM_011527166.2 | XP_011525468.1 | 6 | 1671 | |||
XM_011527167.2 | XP_011525469.1 | 7 | 791 | |||
XM_011527169.2 | XP_011525471.1 | 5 | 590 | |||
XM_011527170.1 | XP_011525472.1 | 5 | 590 | |||
XM_017027039.1 | XP_016882528.1 | 5 | 547 | |||
XM_017027040.1 | XP_016882529.1 | 5 | 4239 | |||
XM_017027041.1 | XP_016882530.1 | 6 | 3327 | |||
XM_017027042.1 | XP_016882531.1 | 7 | 825 | |||
XM_017027043.1 | XP_016882532.1 | 6 | 690 | |||
XM_017027044.1 | XP_016882533.1 | 6 | 732 | |||
XM_017027045.1 | XP_016882534.1 | 6 | 3327 | |||
GenBank mRNA | AK025110.1 | - | 6 | 685 | ||
DA111996.1 | - | 4 | 518 |