Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018110.3 | NP_060580.2 | 4 | 592 | ||
XM_005256042.3 | XP_005256099.1 | 3 | 534 | |||
XM_005256043.2 | XP_005256100.1 | 3 | 534 | |||
GenBank mRNA | AF466369.1 | - | 3 | 242 | ||
AK001350.1 | - | 4 | 519 | |||
AK315426.1 | - | 3 | 502 | |||
BC001540.2 | - | 3 | 291 | 2988350 | ||
BC003541.1 | - | 4 | 534 | 3611472 |