Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021217.2 | NP_067040.1 | 4 | 554 | ||
XM_017027081.1 | XP_016882570.1 | 3 | 417 | |||
XM_017027082.1 | XP_016882571.1 | 4 | 579 | |||
GenBank mRNA | AK290979.1 | - | 4 | 490 | ||
BC043354.1 | - | 4 | 515 | 6014754 | ||
BC062561.1 | - | 4 | 516 | 6644579 | ||
X65230.1 | - | 2 | 146 |