Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BACH2 SIRNA Assays ›
Gene Symbol:
BACH2
Gene Name:
BTB domain and CNC homolog 2
Gene Aliases:
BTBD25
Chromosome Location:
Chr.6: 89926528 - 90296912 on Build GRCh38
UniGene ID:
Hs.269764
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
116313
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BACH2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170794.1
NP_001164265.1
5
1416
NM_021813.3
NP_068585.1
7
1587
XM_005248758.4
XP_005248815.1
4
2369
XM_005248759.4
XP_005248816.1
3
2289
XM_011536039.2
XP_011534341.1
6
1498
XM_011536040.2
XP_011534342.1
5
1447
XM_017011156.1
XP_016866645.1
10
2559
XM_017011157.1
XP_016866646.1
9
2365
XM_017011158.1
XP_016866647.1
11
2638
XM_017011159.1
XP_016866648.1
12
2727
XM_017011160.1
XP_016866649.1
9
2481
XM_017011161.1
XP_016866650.1
11
2189
XM_017011162.1
XP_016866651.1
8
2388
XM_017011163.1
XP_016866652.1
10
2560
XM_017011164.1
XP_016866653.1
9
2466
XM_017011165.1
XP_016866654.1
6
1513
XM_017011166.1
XP_016866655.1
8
1671
XM_017011167.1
XP_016866656.1
10
2559
GenBank mRNA
AB208889.1
-
5
1053
AF357835.1
-
4
912
AJ271878.1
-
7
1421
AK309526.1
-
7
1718
HE578167.1
-
1
460
HE578168.1
-
1
462
HE578169.1
-
1
466
HE578170.1
-
1
462
HE578171.1
-
1
463
HE578172.1
-
1
462
HE578173.1
-
1
463
HE578174.1
-
1
462
HE578175.1
-
1
465
HE578176.1
-
1
471
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
protein ubiquitination
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II distal enhancer sequence-specific binding
transcription factor activity, sequence-specific DNA binding
ubiquitin-protein transferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-cp9cp:80/100.66.77.247:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline