Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BACH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170794.1 | NP_001164265.1 | 7 | 4215 | ||
NM_021813.3 | NP_068585.1 | 9 | 4386 | |||
XM_005248758.4 | XP_005248815.1 | 6 | 5168 | |||
XM_005248759.4 | XP_005248816.1 | 5 | 5088 | |||
XM_017011156.1 | XP_016866645.1 | 12 | 5358 | |||
XM_017011157.1 | XP_016866646.1 | 11 | 5164 | |||
XM_017011158.1 | XP_016866647.1 | 13 | 5437 | |||
XM_017011159.1 | XP_016866648.1 | 14 | 5526 | |||
XM_017011160.1 | XP_016866649.1 | 11 | 5280 | |||
XM_017011161.1 | XP_016866650.1 | 13 | 4988 | |||
XM_017011162.1 | XP_016866651.1 | 10 | 5187 | |||
XM_017011163.1 | XP_016866652.1 | 12 | 5359 | |||
XM_017011164.1 | XP_016866653.1 | 11 | 5265 | |||
XM_017011166.1 | XP_016866655.1 | 10 | 4470 | |||
GenBank mRNA | AJ271878.1 | - | 9 | 4218 |