Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF768 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024671.3 | NP_078947.3 | 2 | 1861 | ||
XM_017023665.1 | XP_016879154.1 | 2 | 2866 | |||
XM_017023666.1 | XP_016879155.1 | 2 | 1686 | |||
GenBank mRNA | AK027089.1 | - | 2 | 1633 | ||
AK295274.1 | - | 3 | 1462 | |||
BC013760.1 | - | 2 | 1861 | 3856266 | ||
BC092403.1 | - | 2 | 1773 | 6201079 |