Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF442 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030824.2 | NP_110451.1 | 6 | 922 | ||
XM_006722908.3 | XP_006722971.1 | 5 | 402 | |||
XM_017027316.1 | XP_016882805.1 | 4 | 350 | |||
XM_017027317.1 | XP_016882806.1 | 4 | 252 | |||
XM_017027318.1 | XP_016882807.1 | 4 | 855 | |||
GenBank mRNA | AK024418.1 | - | 6 | 894 | ||
AK295923.1 | - | 3 | 215 |