Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303043.1 | NP_001289972.1 | 3 | 1569 | ||
NM_030935.4 | NP_112197.1 | 3 | 1672 | |||
NR_130118.1 | 3 | 827 | ||||
NR_130119.1 | 3 | 824 | ||||
GenBank mRNA | AJ133115.1 | - | 3 | 1257 | ||
AK296580.1 | - | 3 | 1120 | |||
AK297702.1 | - | 3 | 762 | |||
AK314824.1 | - | 3 | 1467 | |||
BC001486.2 | - | 3 | 1672 | 2966929 | ||
BC001966.1 | - | 3 | 1525 | 3457297 | ||
BC002972.1 | - | 3 | 1579 | 3538175 | ||
BC010406.2 | - | 3 | 1586 | 3832780 | ||
BC031622.1 | - | 3 | 1562 | 5174360 |