Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168378.1 | NP_001161850.1 | 5 | 2258 | ||
NM_001168379.1 | NP_001161851.1 | 5 | 2085 | |||
NM_001243256.1 | NP_001230185.1 | 4 | 1766 | |||
NM_032153.5 | NP_115529.2 | 5 | 2384 | |||
NR_033118.2 | 3 | 1695 | ||||
NR_033119.2 | 3 | 1641 | ||||
NR_040762.1 | 3 | 1542 | ||||
GenBank mRNA | AK056262.1 | - | 5 | 2195 | ||
AK056394.1 | - | 3 | 2569 | |||
AK094286.1 | - | 3 | 1881 | |||
AK124600.1 | - | 1 | 2720 | |||
AK126595.1 | - | 2 | 2921 | |||
AK127733.1 | - | 3 | 1657 | |||
BC126267.1 | - | 5 | 2020 | 8991983 | ||
BC136339.1 | - | 5 | 2020 | 9020326 |