Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCGF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011663.1 | NP_001011663.1 | 10 | 1625 | ||
NM_032154.3 | NP_115530.2 | 7 | 1400 | |||
GenBank mRNA | AB047006.1 | - | 10 | 1579 | ||
AK027885.1 | - | 10 | 1601 | |||
AK290679.1 | - | 10 | 1579 | |||
BC007602.1 | - | 7 | 1351 | 3350074 | ||
BC010235.1 | - | 10 | 1631 | 3461513 |