Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172632.1 | NP_001166103.1 | 1, 2 | 198 | ||
NM_001172633.1 | NP_001166104.1 | 1, 2 | 198 | |||
NM_002543.3 | NP_002534.1 | 1, 2 | 198 | |||
GenBank mRNA | AB010710.1 | - | 1, 2 | 145 | ||
AB102861.1 | - | 1, 2 | 84 | |||
AF035776.1 | - | 1, 2 | 130 | |||
AK292124.1 | - | 1, 2 | 139 | |||
AK295409.1 | - | 1, 2 | 139 | |||
AK298040.1 | - | 1, 3 | 139 | |||
AK308009.1 | - | 1, 2 | 145 | |||
AL551100.3 | - | 1, 2 | 122 | |||
AX429248.1 | - | 1, 2 | 145 | |||
BC022295.1 | - | 1, 2 | 139 | 4722086 | ||
BP219342.1 | - | 1, 2 | 198 |