Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF514 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318005.1 | NP_001304934.1 | 5 | 3086 | ||
NM_032788.2 | NP_116177.1 | 5 | 2706 | |||
XM_006712806.2 | XP_006712869.1 | 5 | 2517 | |||
GenBank mRNA | AK027363.1 | - | 5 | 2639 | ||
AL832263.1 | - | 3 | 4702 |