Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002556.2 | NP_002547.1 | 2 | 857 | ||
GenBank mRNA | AF185696.1 | - | 2 | 857 | ||
BC011581.1 | - | 2 | 445 | 4560111 | ||
M86917.1 | - | 2 | 857 |