Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002557.3 | NP_002548.3 | 3 | 271 | ||
GenBank mRNA | AB208783.1 | - | 1 | 587 | ||
AK295207.1 | - | 2 | 611 | |||
AK298757.1 | - | 2 | 199 | |||
AK314074.1 | - | 3 | 229 | |||
AY189737.1 | - | 3 | 272 | |||
BC126177.1 | - | 3 | 254 | 8991893 | ||
BC136406.1 | - | 3 | 254 | 9020393 | ||
U09550.1 | - | 3 | 227 |