Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC001610.1 | - | 6 | 1134 | ||
BC017357.1 | - | 6 | 1135 |
Gene Symbol: ZNF331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017026936.1 | XP_016882425.1 | 3 | 734 |