Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135215.1 | NP_001128687.1 | 6 | 1021 | ||
NM_001135216.1 | NP_001128688.1 | 2 | 517 | |||
NM_001243241.1 | NP_001230170.1 | 2 | 442 | |||
NM_030899.4 | NP_112161.3 | 2 | 506 | |||
NM_145909.2 | NP_665916.1 | 6 | 643 | |||
XM_005249295.1 | XP_005249352.1 | 2 | 368 | |||
XM_005249296.4 | XP_005249353.1 | 3 | 4643 | |||
XM_011514807.2 | XP_011513109.1 | 4 | 4693 | |||
XM_011514808.2 | XP_011513110.1 | 2 | 4489 | |||
XM_011514809.1 | XP_011513111.1 | 2 | 353 | |||
XM_011514811.2 | XP_011513113.1 | 2 | 868 | |||
XM_011514812.2 | XP_011513114.1 | 2 | 394 | |||
XM_011514813.2 | XP_011513115.1 | 2 | 857 | |||
XM_017011196.1 | XP_016866685.1 | 4 | 4781 | |||
GenBank mRNA | AF513019.1 | - | 6 | 643 | ||
AK057702.1 | - | 6 | 643 | |||
AK095261.1 | - | 6 | 1021 | |||
AL833617.1 | - | 2 | 518 | |||
BC008490.1 | - | 2 | 280 | 4248596 | ||
DN995648.1 | - | 2 | 190 |