Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145914.2 | NP_666019.1 | 3 | 655 | ||
XM_005250568.4 | XP_005250625.1 | 4 | 1797 | |||
XM_006716113.3 | XP_006716176.1 | 3 | 1636 | |||
XM_017012584.1 | XP_016868073.1 | 4 | 1761 | |||
XM_017012585.1 | XP_016868074.1 | 3 | 679 | |||
GenBank mRNA | AF155100.1 | - | 2 | 330 | ||
AK075324.1 | - | 4 | 755 | |||
AK292267.1 | - | 3 | 675 | |||
AK301871.1 | - | 5 | 706 | |||
AK313884.1 | - | 3 | 673 | |||
AL136865.1 | - | 2 | 608 | |||
BC047309.1 | - | 3 | 648 | 5111890 | ||
BI764964.1 | - | 3 | 654 | |||
CR749301.1 | - | 4 | 539 |