Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152267.3 | NP_689480.2 | 4 | 414 | ||
NR_024210.1 | 4 | 414 | ||||
NR_024212.1 | 3 | 190 | ||||
GenBank mRNA | AK027035.1 | - | 3 | 113 | ||
AK291236.1 | - | 4 | 323 | |||
BC009504.2 | - | 4 | 363 | 2964524 | ||
BC012817.1 | - | 4 | 372 | 2964524 | ||
BC026040.1 | - | 4 | 306 | 4866294 | ||
BC033166.1 | - | 4 | 349 | 4549320 | ||
BC035684.1 | - | 4 | 373 | 5750180 | ||
CR456349.1 | - | 4 | 302 | |||
DB134817.1 | - | 4 | 414 | |||
DQ296559.1 | - | 4 | 355 | |||
DQ296560.1 | - | 4 | 355 | |||
DQ296561.1 | - | 4 | 355 | |||
DQ296563.1 | - | 4 | 355 | |||
DQ296564.1 | - | 4 | 355 | |||
DQ296565.1 | - | 4 | 355 |