Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135825.1 | NP_001129297.1 | 5 | 1438 | ||
NM_152267.3 | NP_689480.2 | 7 | 1606 | |||
NR_024209.1 | 7 | 1591 | ||||
NR_024210.1 | 7 | 1773 | ||||
NR_024211.1 | 6 | 1493 | ||||
NR_024212.1 | 6 | 1382 | ||||
GenBank mRNA | AK027035.1 | - | 6 | 1304 | ||
AK075154.1 | - | 1 | 688 | |||
AK095947.1 | - | 5 | 1347 | |||
BC009504.2 | - | 8 | 972 | 2964524 | ||
BC012817.1 | - | 8 | 981 | 2964524 | ||
BC017302.1 | - | 1 | 502 | |||
BC026040.1 | - | 7 | 1498 | 4866294 | ||
BC033166.1 | - | 7 | 1541 | 4549320 | ||
BC035684.1 | - | 7 | 1565 | 5750180 | ||
BX648019.1 | - | 7 | 1515 | |||
DQ296559.1 | - | 7 | 1547 | |||
DQ296561.1 | - | 8 | 1137 | |||
DQ296562.1 | - | 5 | 1379 | |||
DQ296564.1 | - | 7 | 1715 | |||
DQ296565.1 | - | 7 | 1547 |