Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102651.1 | NP_001096121.1 | 4, 5 | 639 | ||
XM_006722635.3 | XP_006722698.1 | 3, 4 | 326 | |||
XM_011527660.2 | XP_011525962.1 | 4, 5 | 480 | |||
XM_011527662.2 | XP_011525964.1 | 3, 4 | 382 | |||
XM_011527664.2 | XP_011525966.1 | 2, 3 | 1611 | |||
XM_017026234.1 | XP_016881723.1 | 4, 5 | 553 | |||
XM_017026235.1 | XP_016881724.1 | 2, 3 | 1956 | |||
XM_017026236.1 | XP_016881725.1 | 2, 3 | 324 | |||
GenBank mRNA | AK027860.1 | - | 1, 2 | 1935 | ||
AK092136.1 | - | 5, 6 | 691 | |||
BC000113.1 | - | 4, 5 | 550 | |||
BC047777.1 | - | 4, 5 | 608 | 6187000 |