Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM237 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044385.2 | NP_001037850.1 | 5 | 335 | ||
NM_152388.3 | NP_689601.2 | 5 | 500 | |||
GenBank mRNA | AB053301.1 | - | 5 | 502 | ||
AK090601.1 | - | 5 | 193 | |||
AK303954.1 | - | 5 | 307 | |||
BC029611.1 | - | 5 | 248 | 5304046 | ||
BP226834.1 | - | 5 | 335 | |||
BQ219059.1 | - | 5 | 307 |