Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF579 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152600.2 | NP_689813.2 | 2 | 2078 | ||
XM_017026409.1 | XP_016881898.1 | 3 | 1786 | |||
XM_017026410.1 | XP_016881899.1 | 3 | 1786 | |||
XM_017026411.1 | XP_016881900.1 | 3 | 1786 | |||
GenBank mRNA | AK092772.1 | - | 2 | 2054 | ||
BC042926.1 | - | 1 | 916 |