Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042413.1 | NP_001035878.1 | 3 | 1286 | ||
NM_152629.3 | NP_689842.3 | 2 | 302 | |||
XM_005251386.4 | XP_005251443.1 | 2 | 845 | |||
XM_005251389.4 | XP_005251446.1 | 3 | 1335 | |||
XM_006716731.3 | XP_006716794.1 | 3 | 1335 | |||
XM_011517763.2 | XP_011516065.1 | 3 | 965 | |||
XM_011517764.2 | XP_011516066.1 | 3 | 795 | |||
XM_011517765.2 | XP_011516067.1 | 3 | 1332 | |||
XM_011517766.2 | XP_011516068.1 | 2 | 1374 | |||
XM_011517769.2 | XP_011516071.1 | 3 | 1335 | |||
XM_017014361.1 | XP_016869850.1 | 2 | 1073 | |||
GenBank mRNA | BC033899.2 | - | 2 | 302 | 4828458 | |
DQ438877.1 | - | 1167 | ||||
DQ438886.1 | - | 3 | 1167 | |||
DQ438887.1 | - | 3 | 845 | |||
DQ438888.1 | - | 3 | 784 | |||
DQ438889.1 | - | 2 | 1144 | |||
DQ438890.1 | - | 2 | 483 | |||
DQ438891.1 | - | 2 | 681 | |||
DQ438892.1 | - | 2 | 359 | |||
DQ438893.1 | - | 2 | 533 | |||
DQ438894.1 | - | 2 | 540 | |||
DQ438895.1 | - | 3 | 340 | |||
DQ438896.1 | - | 2 | 298 | |||
DQ438905.1 | - | 2 | 2164 | |||
KU178885.1 | - | 1 | 108 | |||
KU178886.1 | - | 1 | 108 | |||
KU178887.1 | - | 1 | 108 |