Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001214906.1 | NP_001201835.1 | 3 | 1962 | ||
NM_001214907.1 | NP_001201836.1 | 2 | 1866 | |||
NM_001214909.1 | NP_001201838.1 | 3 | 2330 | |||
NM_001324494.1 | NP_001311423.1 | 2 | 2236 | |||
NM_152652.2 | NP_689865.2 | 2 | 2224 | |||
GenBank mRNA | AK056313.1 | - | 2 | 1907 | ||
AK122598.1 | - | 1 | 991 | |||
AL832433.1 | - | 1 | 907 | |||
BC007393.2 | - | 1 | 493 | |||
BC041388.1 | - | 2 | 1911 | 5276303 |