Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF449 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152695.5 | NP_689908.3 | 3 | 686 | ||
XM_011531312.2 | XP_011529614.1 | 4 | 756 | |||
XM_011531313.2 | XP_011529615.1 | 2 | 1302 | |||
XM_011531314.2 | XP_011529616.1 | 4 | 1443 | |||
XM_017029350.1 | XP_016884839.1 | 3 | 634 | |||
XM_017029351.1 | XP_016884840.1 | 3 | 731 | |||
GenBank mRNA | AK314344.1 | - | 3 | 663 | ||
AL833109.1 | - | 3 | 644 | |||
AL833407.1 | - | 3 | 662 | |||
AY280801.1 | - | 3 | 662 | |||
BC060768.1 | - | 3 | 661 | 30346713 | ||
BC065938.1 | - | 4 | 733 |