Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023560.3 | NP_001018854.2 | 4 | 2275 | ||
NM_001111039.2 | NP_001104509.1 | 4 | 2272 | |||
NM_001287421.1 | NP_001274350.1 | 4 | 1761 | |||
NM_001287422.1 | NP_001274351.1 | 2 | 1671 | |||
NM_152736.5 | NP_689949.3 | 4 | 1870 | |||
XM_011514862.2 | XP_011513164.1 | 4 | 2474 | |||
XM_011514864.1 | XP_011513166.1 | 5 | 2051 | |||
XM_011514866.1 | XP_011513168.1 | 4 | 1823 | |||
XM_011514867.1 | XP_011513169.1 | 4 | 2187 | |||
XM_011514869.1 | XP_011513171.1 | 5 | 2115 | |||
XM_017011264.1 | XP_016866753.1 | 4 | 2472 | |||
XM_017011265.1 | XP_016866754.1 | 4 | 2206 | |||
GenBank mRNA | AJ420533.1 | - | 1 | 607 | ||
AK095948.1 | - | 4 | 1822 | |||
AL832741.1 | - | 3 | 2463 | |||
BC013951.2 | - | 4 | 1726 | 2963683 | ||
BC013962.2 | - | 4 | 1726 | 2963683 | ||
BX537535.1 | - | 4 | 2144 |