Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UHRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152896.2 | NP_690856.1 | 3 | 885 | ||
NR_046386.1 | 3 | 885 | ||||
XM_011517704.2 | XP_011516006.1 | 3 | 514 | |||
XM_017014253.1 | XP_016869742.1 | 3 | 888 | |||
XM_017014254.1 | XP_016869743.1 | 3 | 514 | |||
GenBank mRNA | AB071698.1 | - | 3 | 583 | ||
AF274049.1 | - | 3 | 713 | |||
AK308433.1 | - | 3 | 837 | |||
BC028397.2 | - | 3 | 885 | 4797830 |