Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278623.1 | NP_001265552.1 | 2, 3 | 298 | ||
NM_001278624.1 | NP_001265553.1 | 2, 3 | 436 | |||
NM_152995.5 | NP_694540.3 | 2, 3 | 338 | |||
NR_103795.1 | 2, 3 | 298 | ||||
GenBank mRNA | AB085695.1 | - | 2, 3 | 294 | ||
AB181916.1 | - | 2, 3 | 418 | |||
AK302440.1 | - | 2, 3 | 296 | |||
BC051193.1 | - | 2, 3 | 269 | 5271401 | ||
DB087408.1 | - | 2, 3 | 339 |