Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002597.4 | NP_002588.3 | 3 | 223 | ||
NM_022576.3 | NP_072098.1 | 2 | 185 | |||
XM_011509603.2 | XP_011507905.1 | 2 | 1241 | |||
GenBank mRNA | AF076464.1 | - | 2 | 186 | ||
BC093839.1 | - | 1 | 16 | |||
BC112134.1 | - | 1 | 16 | |||
EL945964.1 | - | 3 | 227 | |||
M33478.1 | - | 3 | 200 |