Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153018.2 | NP_694563.1 | 2 | 1513 | ||
GenBank mRNA | AK055288.1 | - | 2 | 1513 | ||
AK125731.1 | - | 1 | 760 | |||
AK290549.1 | - | 2 | 1505 | |||
BC142962.1 | - | 2 | 1492 | 8860374 | ||
BX647638.1 | - | 2 | 1466 |