Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143941.1 | NP_001137413.1 | 2 | 264 | ||
NM_001143942.1 | NP_001137414.1 | 2 | 280 | |||
NM_153020.2 | NP_694565.1 | 1 | 212 | |||
XM_005248932.3 | XP_005248989.1 | 2 | 350 | |||
XM_011514387.2 | XP_011512689.1 | 2 | 209 | |||
GenBank mRNA | AK055391.1 | - | 1 | 212 | ||
AK095016.1 | - | 2 | 315 | |||
AK129865.1 | - | 2 | 153 | |||
AK293052.1 | - | 2 | 280 | |||
AK315856.1 | - | 2 | 280 | |||
AY547318.1 | - | 2 | 276 | |||
BC104808.1 | - | 1 | 180 | 8143811 | ||
BC104810.1 | - | 1 | 180 | 8143813 | ||
BP309176.1 | - | 2 | 264 |