Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCORL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153686.8 | NP_710153.2 | 7 | 1046 | ||
NR_136669.1 | 5 | 872 | ||||
XM_011513821.2 | XP_011512123.1 | 9 | 5777 | |||
XM_011513822.2 | XP_011512124.1 | 8 | 5750 | |||
XM_011513823.2 | XP_011512125.1 | 8 | 5623 | |||
XM_011513826.2 | XP_011512128.1 | 6 | 830 | |||
XM_017007961.1 | XP_016863450.1 | 8 | 7059 | |||
XM_017007962.1 | XP_016863451.1 | 8 | 5701 | |||
XM_017007963.1 | XP_016863452.1 | 8 | 6004 | |||
XM_017007965.1 | XP_016863454.1 | 6 | 2105 | |||
GenBank mRNA | AK096181.1 | - | 8 | 907 | ||
AK299939.1 | - | 5 | 754 | |||
BC037322.1 | - | 7 | 929 | 5260413 |